missing translation for 'onlineSavingsMsg'
Learn More

p63/TP73L Antibody (TP63/11), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-07431-20ug

 View more versions of this product

Product Code. 18665186

  • £218.00 / 20µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

p63/TP73L Monoclonal antibody specifically detects p63/TP73L in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

p63/TP73L
Monoclonal
0.2 mg/mL
Immunohistochemistry-Paraffin 0.25-0.5 μg/mL
Q9H3D4, Q9H3D4
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse
Purified
Immunohistochemistry (Paraffin)
TP63/11
Unconjugated
10 mM PBS with 0.05% BSA
amplified in squamous cell carcinoma, Chronic μgcerative stomatitis protein, CUSP, EEC3B(p51B), Keratinocyte transcription factor KET, KETRHS, LMS, OFC8SHFM4TP73LNBP, p40, p51tumor protein p53-like, p53CP, P63, p63AIS, p73H, P73L, p73LB(p51A), TP53CP, TP53L, Transformation-related protein 63, tumor protein 63, tumor protein p53-competing protein, tumor protein p63, tumor protein p63 deltaN isoform delta, Tumor protein p73-like
Recombinant full-length human p63/TP73L protein (Uniprot: Q9H3D4)
20 μg
Apoptosis, Cancer, Cellular Markers, Hypoxia, Stem Cell Markers, Tumor Suppressors
8626
Store at 4°C.
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.