missing translation for 'onlineSavingsMsg'
Learn More

p63 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £375.00

Specifications

Antigen p63
Applications Immunofluorescence, Flow Cytometry (Intracellular Staining)
Classification Polyclonal
Conjugate CoraLite 488
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17781249
View Documents
Proteintech
CL488-12143-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16850566
View Documents
Proteintech
CL488-12143-100UL
100 μL
£375.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The p63 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human p63. This antibody recognizes human, mouse, rat antigen. The p63 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.

Specifications

Specifications

p63
Polyclonal
Rabbit
8626
Primary
Immunofluorescence, Flow Cytometry (Intracellular Staining)
CoraLite 488
Human, Rat, Mouse
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.