Learn More
p63 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-12143-20UL
 View more versions of this product
Product Code. 17781249
  • £135.00 / 20µL
Description
The p63 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human p63. This antibody recognizes human, mouse, rat antigen. The p63 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifications
Specifications
p63
Polyclonal
CoraLite 488
20 μL
8626
IgG
Immunofluorescence, Flow Cytometry (Intracellular Staining)
1200 ug/mL
Rabbit
Primary
Human, Rat, Mouse