missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p53 DINP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£167.00 - £302.00
Specifications
| Antigen | p53 DINP1 |
|---|---|
| Dilution | Western Blot 1:1000-1:5000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18066845
|
Novus Biologicals
NB100-56627SS |
0.05 mL |
£167.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18060544
|
Novus Biologicals
NB100-56627 |
0.2 mL |
£302.00
0.20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p53 DINP1 Polyclonal specifically detects p53 DINP1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| p53 DINP1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, DNA Double Strand Break Repair | |
| Whole antisera with PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
| DKFZp434M1317, FLJ22139, p53DINP1, P53DINP1p53-dependent damage-inducible nuclear protein 1, p53-inducible p53DINP1, SIPStress-induced protein, Teap, TP53DINP1, TP53INP1A, TP53INP1B, tumor protein p53 inducible nuclear protein 1, tumor protein p53-inducible nuclear protein 1 | |
| TP53INP1 | |
| IgG | |
| Unpurified |
| Western Blot 1:1000-1:5000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat | |
| Q96A56 | |
| 94241 | |
| This antibody was generated by immunizing rabbit with synthetic peptides of human p53DINP1. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title