missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p53 DINP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56627
This item is not returnable.
View return policy
Description
p53 DINP1 Polyclonal specifically detects p53 DINP1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| p53 DINP1 | |
| Polyclonal | |
| Western Blot 1:1000-1:5000 | |
| Q96A56 | |
| TP53INP1 | |
| This antibody was generated by immunizing rabbit with synthetic peptides of human p53DINP1. | |
| 0.2 mL | |
| Apoptosis, DNA Double Strand Break Repair | |
| 94241 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Whole antisera with PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
| DKFZp434M1317, FLJ22139, p53DINP1, P53DINP1p53-dependent damage-inducible nuclear protein 1, p53-inducible p53DINP1, SIPStress-induced protein, Teap, TP53DINP1, TP53INP1A, TP53INP1B, tumor protein p53 inducible nuclear protein 1, tumor protein p53-inducible nuclear protein 1 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction