missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p53 AIP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76742-0.025mg
This item is not returnable.
View return policy
Description
p53 AIP1 Polyclonal specifically detects p53 AIP1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry.
Specifications
| p53 AIP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| p53AIP1, p53-regulated apoptosis-inducing protein 1, tumor protein p53 regulated apoptosis inducing protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 4-8 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 μg/mL | |
| BAB16421 | |
| TP53AIP1 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human p53AIP1. The immunogen is located within the first 50 amino acids of p53AIP1. Amino Acid Squence: GSSSEASFRSAQASC | |
| 0.025 mg | |
| Apoptosis, DNA Repair | |
| 63970 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction