missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p53 AIP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£133.00
Specifications
| Antigen | p53 AIP1 |
|---|---|
| Concentration | 1 mg/ml |
| Dilution | Western Blot 4-8 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
p53 AIP1 Polyclonal specifically detects p53 AIP1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry.Specifications
| p53 AIP1 | |
| Western Blot 4-8 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, DNA Repair | |
| PBS with 0.02% Sodium Azide | |
| p53AIP1, p53-regulated apoptosis-inducing protein 1, tumor protein p53 regulated apoptosis inducing protein 1 | |
| TP53AIP1 | |
| IgG | |
| Affinity Purified |
| 1 mg/ml | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| BAB16421 | |
| 63970 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human p53AIP1. The immunogen is located within the first 50 amino acids of p53AIP1. Amino Acid Squence: GSSSEASFRSAQASC | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title