missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p38 beta/MAPK11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-56446SS
This item is not returnable.
View return policy
Description
p38 beta/MAPK11 Polyclonal specifically detects p38 beta/MAPK11 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| p38 beta/MAPK11 | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
| EC 2.7.11, EC 2.7.11.24, MAP kinase 11, MAP kinase p38 beta, MAPK 11, mitogen-activated protein kinase 11, Mitogen-activated protein kinase p38 beta, mitogen-activated protein kinase p38-2, p38b, p38Beta, P38BETA2, PRKM11, SAPK2B, SAPK2p38-2P38B, Stress-activated protein kinase 2, stress-activated protein kinase-2, stress-activated protein kinase-2b | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| In human or mouse brain, a 38 kDa band is observed. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 1.0 mg/mL | |
| Western Blot 1-3 ug/ml | |
| NP_002742 | |
| MAPK11 | |
| A synthetic peptide corresponding to a portion amino acids 1-100 was used as immunogen, | |
| 0.025 mg | |
| Breast Cancer, Cancer, Protein Kinase | |
| 5600 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction