missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p38 beta/MAPK11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£177.00 - £319.00
Specifications
| Antigen | p38 beta/MAPK11 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-3 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18014595
|
Novus Biologicals
NB100-56446SS |
0.025 mg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18700374
|
Novus Biologicals
NB100-56446 |
£319.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
p38 beta/MAPK11 Polyclonal specifically detects p38 beta/MAPK11 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| p38 beta/MAPK11 | |
| Western Blot 1-3 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| NP_002742 | |
| 5600 | |
| A synthetic peptide corresponding to a portion amino acids 1-100 was used as immunogen, | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Protein Kinase | |
| PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
| EC 2.7.11, EC 2.7.11.24, MAP kinase 11, MAP kinase p38 beta, MAPK 11, mitogen-activated protein kinase 11, Mitogen-activated protein kinase p38 beta, mitogen-activated protein kinase p38-2, p38b, p38Beta, P38BETA2, PRKM11, SAPK2B, SAPK2p38-2P38B, Stress-activated protein kinase 2, stress-activated protein kinase-2, stress-activated protein kinase-2b | |
| MAPK11 | |
| IgG | |
| Protein G purified | |
| In human or mouse brain, a 38 kDa band is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title