missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p38 alpha Antibody (9F12), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
£155.00
Specifications
| Antigen | p38 |
|---|---|
| Clone | 9F12 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:1000, Proximity Ligation Assay |
| Applications | Western Blot |
Description
p38 alpha Monoclonal specifically detects p38 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Proximity Ligation Assay, Microarray.Specifications
| p38 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| CSAID-binding protein, Csaids binding protein, CSBP, CSBP1MAX-interacting protein 2, CSBP2, CSPB1, cytokine suppressive inflammatory drug binding protein, Cytokine suppressive inflammatory drug-binding protein, EC 2.7.11, EC 2.7.11.24, EXIP, MAP kinase 14, MAP kinase MXI2, MAP kinase p38 alpha, MAPK 14, mitogen-activated protein kinase 14, Mitogen-activated protein kinase p38 alpha, Mxi2, p38, p38 MAP kinase, p38 mitogen activated protein kinase, p38ALPHA, p38alpha Exip, PRKM14, PRKM15, RK, SAPK2A, stress-activated protein kinase 2A | |
| MAPK14 | |
| IgG1 | |
| Protein G purified | |
| Detects approx 38kDa. |
| 9F12 | |
| Western Blot 1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:1000, Proximity Ligation Assay | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q16539, NP_001306.1 | |
| 1432 | |
| Full length recombinant protein expressed in E.coli cells | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title