missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
p38 alpha Monoclonal specifically detects p38 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Proximity Ligation Assay, Microarray.
Specifications
Specifications
| Antigen | p38 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 9F12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:1000, Proximity Ligation Assay |
| Gene Accession No. | Q16539, NP_001306.1 |
| Gene Alias | CSAID-binding protein, Csaids binding protein, CSBP, CSBP1MAX-interacting protein 2, CSBP2, CSPB1, cytokine suppressive inflammatory drug binding protein, Cytokine suppressive inflammatory drug-binding protein, EC 2.7.11, EC 2.7.11.24, EXIP, MAP kinase 14, MAP kinase MXI2, MAP kinase p38 alpha, MAPK 14, mitogen-activated protein kinase 14, Mitogen-activated protein kinase p38 alpha, Mxi2, p38, p38 MAP kinase, p38 mitogen activated protein kinase, p38ALPHA, p38alpha Exip, PRKM14, PRKM15, RK, SAPK2A, stress-activated protein kinase 2A |
| Gene Symbols | MAPK14 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?