missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p35 baculovirus Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 9 publications
£205.00 - £345.00
Specifications
| Antigen | p35 baculovirus |
|---|---|
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 21514385), Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen Assay Dependent, Immunocytochemistry |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18058465
|
Novus Biologicals
NB100-56153SS |
0.025 mL |
£205.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18710474
|
Novus Biologicals
NB100-56153 |
0.05 mL |
£345.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p35 baculovirus Polyclonal specifically detects p35 baculovirus in Drosophila, Silkworm, Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunocytochemistry.Specifications
| p35 baculovirus | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Signal Transduction | |
| Neat whole antisera with 0.05% Sodium Azide | |
| Full-length recombinant p35 baculovirus protein was used as immunogen. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| 34 kDa |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 21514385), Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen Assay Dependent, Immunocytochemistry | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Drosophila, Silkworm, Virus | |
| p35 Baculovirus, p35-Baculovirus | |
| IgG | |
| Unpurified | |
| p35 baculovirus may be observed as one or multiple bands on western blot: p35 baculovirus is cleaved during its reaction with caspases. This results in cleavage of p35; 10 and 25 kDa fragments have been described. Additional breakdown p35 products may also result. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title