missing translation for 'onlineSavingsMsg'
Learn More

p35 baculovirus Antibody, Novus Biologicals™

Product Code. 18058465 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
Unit Size:
0.05mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18058465 0.025 mL 25µL
18710474 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18058465 Supplier Novus Biologicals Supplier No. NB10056153SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 9 publications

p35 baculovirus Polyclonal specifically detects p35 baculovirus in Drosophila, Silkworm, Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p35 baculovirus
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 21514385), Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen Assay Dependent, Immunocytochemistry
Formulation Neat whole antisera with 0.05% Sodium Azide
Gene Alias p35 Baculovirus, p35-Baculovirus
Host Species Rabbit
Immunogen Full-length recombinant p35 baculovirus protein was used as immunogen.
Molecular Weight of Antigen 34 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Test Specificity p35 baculovirus may be observed as one or multiple bands on western blot: p35 baculovirus is cleaved during its reaction with caspases. This results in cleavage of p35; 10 and 25 kDa fragments have been described. Additional breakdown p35 products may also result.
Target Species Drosophila, Silkworm, Virus
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.