missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y6/P2RY6 Rabbit anti-Human, Clone: 4I8C9, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £396.00
Specifications
| Antigen | P2Y6/P2RY6 |
|---|---|
| Clone | 4I8C9 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18074835
|
Novus Biologicals
NBP3-16259-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18307855
|
Novus Biologicals
NBP3-16259-100UL |
100 μg |
£396.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
P2Y6/P2RY6 Monoclonal antibody specifically detects P2Y6/P2RY6 in Human samples. It is validated for Western BlotSpecifications
| P2Y6/P2RY6 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cytokine Research, GPCR, Plasma Membrane Markers | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 5031 | |
| IgG | |
| Affinity purified |
| 4I8C9 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| G-coupled nucleotide receptor, MGC15335, P2 purinoceptor, P2Y6 receptor, P2Y6P2Y purinoceptor 6, pyrimidinergic receptor P2Y, G-protein coupled, 6 | |
| A synthetic peptide corresponding to a sequence within amino acids 229-328 of human P2Y6/P2RY6 (Q15077). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title