missing translation for 'onlineSavingsMsg'
Learn More

P2Y6/P2RY6 Rabbit anti-Human, Clone: 4I8C9, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-16259-100UL

 View more versions of this product

Product Code. 18307855

  • £396.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

P2Y6/P2RY6 Monoclonal antibody specifically detects P2Y6/P2RY6 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

P2Y6/P2RY6
Monoclonal
Unconjugated
G-coupled nucleotide receptor, MGC15335, P2 purinoceptor, P2Y6 receptor, P2Y6P2Y purinoceptor 6, pyrimidinergic receptor P2Y, G-protein coupled, 6
A synthetic peptide corresponding to a sequence within amino acids 229-328 of human P2Y6/P2RY6 (Q15077).
100 μg
Apoptosis, Cytokine Research, GPCR, Plasma Membrane Markers
5031
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot
4I8C9
PBS, 0.05% BSA, 50% glycerol, pH7.3
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.