missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y4/P2RY4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95193-0.1ml
This item is not returnable.
View return policy
Description
P2Y4/P2RY4 Polyclonal antibody specifically detects P2Y4/P2RY4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| P2Y4/P2RY4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| P2P, P2Y4P2Y purinoceptor 4, pyrimidinergic receptor P2Y, G-protein coupled, 4, UNRNRUUridine nucleotide receptor | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 286-365 of human P2RY4 (NP_002556.1). VVYKVTRPLASANSCLDPVLYLLTGDKYRRQLRQLCGGGKPQPRTAASSLALVSLPEDSSCRWAATPQDSSCSTPRADRL | |
| 0.1 mL | |
| GPCR | |
| 5030 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction