missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2X6/P2RX6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | P2X6/P2RX6 |
|---|---|
| Dilution | Western Blot 1:200-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18688231
|
Novus Biologicals
NBP2-93169-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18687261
|
Novus Biologicals
NBP2-93169-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
P2X6/P2RX6 Polyclonal antibody specifically detects P2X6/P2RX6 in Mouse samples. It is validated for Western BlotSpecifications
| P2X6/P2RX6 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cardiovascular Biology, Endocrinology, Neuroscience, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 9127 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| ATP receptor, MGC129625, P2RXL1skeletal muscle-expressed P2X, P2X purinoceptor 6, P2X6purinoceptor P2X6, P2XMpurinoreceptor P2X6, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 6, Purinergic receptor P2X-like 1, purinergic receptor P2X-like 1, orphan receptor | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-280 of human P2RX6 (NP_001153026.1). WGLLDYKTEKWALLAKKGYQERDLEPQFSIITKLKGVSVTQIKELGNRLWDVADFVKPPQGENVFFLVTNFLVTPAQVQGRCPEHPSVPLANCWVDEDCPEGEGGTHSHGVKTGQCVVFNGTHRTCEIWSWCPVESGVVPSRPLLAQAQNFTLFIKNTVTFSKFNFSKSNALETWDPTYFKHCRYEPQFSPYCPVFRIGDLVAKAGGTFEDLALLGGSVGIRVHWDCDLDTGDSGCWPHYSFQLQEKSYNFRTATHWWEQP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title