missing translation for 'onlineSavingsMsg'
Learn More

P2X6/P2RX6 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-93169-0.1ml

 View more versions of this product

Product Code. 18687261

  • £366.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

P2X6/P2RX6 Polyclonal antibody specifically detects P2X6/P2RX6 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

P2X6/P2RX6
Polyclonal
Western Blot 1:200-1:1000
ATP receptor, MGC129625, P2RXL1skeletal muscle-expressed P2X, P2X purinoceptor 6, P2X6purinoceptor P2X6, P2XMpurinoreceptor P2X6, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 6, Purinergic receptor P2X-like 1, purinergic receptor P2X-like 1, orphan receptor
Recombinant fusion protein containing a sequence corresponding to amino acids 20-280 of human P2RX6 (NP_001153026.1). WGLLDYKTEKWALLAKKGYQERDLEPQFSIITKLKGVSVTQIKELGNRLWDVADFVKPPQGENVFFLVTNFLVTPAQVQGRCPEHPSVPLANCWVDEDCPEGEGGTHSHGVKTGQCVVFNGTHRTCEIWSWCPVESGVVPSRPLLAQAQNFTLFIKNTVTFSKFNFSKSNALETWDPTYFKHCRYEPQFSPYCPVFRIGDLVAKAGGTFEDLALLGGSVGIRVHWDCDLDTGDSGCWPHYSFQLQEKSYNFRTATHWWEQP
0.1 mL
Cancer, Cardiovascular Biology, Endocrinology, Neuroscience, Signal Transduction
9127
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.