missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2X3/P2RX3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
£153.00 - £397.00
Specifications
| Antigen | P2X3/P2RX3 |
|---|---|
| Concentration | LYOPH |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18142945
|
Novus Biologicals
NBP2-14838-0.025ML |
0.025 mL |
£153.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18170662
|
Novus Biologicals
NBP2-14838 |
0.1 mL |
£397.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
P2X3/P2RX3 Polyclonal specifically detects P2X3/P2RX3 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| P2X3/P2RX3 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Goat | |
| Rat, Mouse | |
| 5024 | |
| IgG | |
| Unpurified |
| LYOPH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| ATP receptor, MGC129956, P2X purinoceptor 3, P2X receptor, subunit 3, P2X3purinergic receptor P2X3, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 3, purinoceptor P2X3 | |
| P2RX3 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title