missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2X3/P2RX3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-14838
This item is not returnable.
View return policy
Description
P2X3/P2RX3 Polyclonal specifically detects P2X3/P2RX3 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| P2X3/P2RX3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from whole antisera with No Preservative | |
| P2RX3 | |
| A synthetic peptide from the cytoplasmic domain of mouse P2X3/P2RX3 conjugated to blue carrier protein was used as the antigen. The peptide is homologous in rat. | |
| 0.1 mL | |
| Primary | |
| Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunohistochemistry-Paraffin 1:300-1:2000 | |
| ATP receptor, MGC129956, P2X purinoceptor 3, P2X receptor, subunit 3, P2X3purinergic receptor P2X3, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 3, purinoceptor P2X3 | |
| Goat | |
| Unpurified | |
| RUO | |
| 5024 | |
| Mouse, Rat | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction