missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p19ARF/CDKN2A Antibody (5-C3-1) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£137.00 - £372.00
Specifications
| Antigen | p19ARF |
|---|---|
| Clone | 5-C3-1 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1 ug/mL, Immunohistochemistry 5 u/gml, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 27177758), Immunohistochemistry-Frozen 5 ug/ml, Flow (Intracellular), CyTOF-ready, Knockout Validated |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18737223
|
Novus Biologicals
NB200-174SS |
0.025 mL |
£137.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18701154
|
Novus Biologicals
NB200-174 |
0.1 mL |
£372.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p19ARF/CDKN2A Monoclonal specifically detects p19ARF/CDKN2A in Human, Mouse, Golden Syrian Hamster (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready, Knockout Validated.Specifications
| p19ARF | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rat | |
| ABC Transporters, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, Hypoxia, MAP Kinase Signaling, Neuroscience, p53 Pathway, Transcription Factors and Regulators, Tumor Suppressors | |
| ARF, CDK4 inhibitor p16-INK4, CDK4IP14ARF, CDKN2, cell cycle negative regulator beta, CMM2P16-INK4A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4), INK4, INK4A, MLMP16INK4, MTS-1, MTS1P14, Multiple tumor suppressor 1, p14, p14ARF, p16, p16-INK4, p16INK4a, p16-INK4a, P19, p19ARF, TP16 | |
| CDKN2A | |
| IgG2b | |
| Protein A purified | |
| Mouse. The antibody does not react with human or Golden Syrian hamster. Armenian hamster has not been tested. |
| 5-C3-1 | |
| Western Blot 1:500-1:1000, Flow Cytometry 1 ug/mL, Immunohistochemistry 5 u/gml, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 27177758), Immunohistochemistry-Frozen 5 ug/ml, Flow (Intracellular), CyTOF-ready, Knockout Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Golden Syrian Hamster (Negative) | |
| 12578 | |
| A synthetic peptide containing amino acids 54-75 of the murine p19ARF. [UniProt# Q64364] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 19 kDa |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto