missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p19ARF/CDKN2A Antibody (5-C3-1) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB200-174
This item is not returnable.
View return policy
Description
p19ARF/CDKN2A Monoclonal specifically detects p19ARF/CDKN2A in Human, Mouse, Golden Syrian Hamster (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready, Knockout Validated.
Specifications
| p19ARF | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, Flow Cytometry 1 ug/mL, Immunohistochemistry 5 u/gml, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 27177758), Immunohistochemistry-Frozen 5 ug/ml, Flow (Intracellular), CyTOF-ready, Knockout Validated | |
| CDKN2A | |
| A synthetic peptide containing amino acids 54-75 of the murine p19ARF. [UniProt# Q64364] | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse. The antibody does not react with human or Golden Syrian hamster. Armenian hamster has not been tested. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 5-C3-1 | |
| Unconjugated | |
| ARF, CDK4 inhibitor p16-INK4, CDK4IP14ARF, CDKN2, cell cycle negative regulator beta, CMM2P16-INK4A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4), INK4, INK4A, MLMP16INK4, MTS-1, MTS1P14, Multiple tumor suppressor 1, p14, p14ARF, p16, p16-INK4, p16INK4a, p16-INK4a, P19, p19ARF, TP16 | |
| Rat | |
| 19 kDa | |
| 0.1 mL | |
| ABC Transporters, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, Hypoxia, MAP Kinase Signaling, Neuroscience, p53 Pathway, Transcription Factors and Regulators, Tumor Suppressors | |
| 12578 | |
| Human, Mouse, Golden Syrian Hamster (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction