missing translation for 'onlineSavingsMsg'
Learn More

p19 INK4d Antibody (06), Novus Biologicals™

Mouse Monoclonal Antibody

£173.00 - £255.00

Specifications

Antigen p19 INK4d
Clone 06
Dilution ELISA 1:1000-1:2000
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18601028
View Documents
Novus Biologicals
NBP3-05845-100ul
100 μg
£255.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18646676
View Documents
Novus Biologicals
NBP3-05845-50ul
50 μg
£173.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

p19 INK4d Monoclonal antibody specifically detects p19 INK4d in Human samples. It is validated for ELISA
Specifications

Specifications

p19 INK4d
ELISA 1:1000-1:2000
Monoclonal
Purified
RUO
Human
CDK inhibitor p19INK4d, cell cycle inhibitor, Nur77 associating protein, cyclin-dependent kinase 4 inhibitor D, cyclin-dependent kinase 4 inhibitor D p19, cyclin-dependent kinase inhibitor 2D (p19, inhibits CDK4), inhibitor of cyclin-dependent kinase 4d, INK4D, p19, p19-INK4D
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human p19 INK4d (Uniprot#: P55273; Met 10-Leu 166).
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
06
ELISA
Unconjugated
Mouse
Cell Cycle and Replication, Ovarian Carcinoma Cell Markers
0.2 um filtered solution in PBS
1032
IgG1
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.