missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p19 INK4d Antibody (06), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05845-100ul
This item is not returnable.
View return policy
Description
p19 INK4d Monoclonal antibody specifically detects p19 INK4d in Human samples. It is validated for ELISA
Specifications
| p19 INK4d | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 06 | |
| ELISA 1:1000-1:2000 | |
| CDK inhibitor p19INK4d, cell cycle inhibitor, Nur77 associating protein, cyclin-dependent kinase 4 inhibitor D, cyclin-dependent kinase 4 inhibitor D p19, cyclin-dependent kinase inhibitor 2D (p19, inhibits CDK4), inhibitor of cyclin-dependent kinase 4d, INK4D, p19, p19-INK4D | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human p19 INK4d (Uniprot#: P55273; Met 10-Leu 166). | |
| 100 μg | |
| Cell Cycle and Replication, Ovarian Carcinoma Cell Markers | |
| 1032 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction