missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p130Cas/BCAR1, p Tyr410 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£154.00 - £539.00
Specifications
| Antigen | p130Cas/BCAR1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18215753
|
Novus Biologicals
NBP2-54638-0.05MG |
0.05 mg |
£345.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611147
|
Novus Biologicals
NBP2-54638 |
0.1 mg |
£539.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620387
|
Novus Biologicals
NBP2-54638-0.025mg |
0.025 mg |
£154.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p130Cas/BCAR1 Polyclonal specifically detects p130Cas/BCAR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| p130Cas/BCAR1 | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100 | |
| Polyclonal | |
| Rabbit | |
| breast cancer estrogen resistance 1, Cas scaffolding protein family member 1, CAS1, CASFLJ45059, CASS1CRKASbreast cancer estrogen resistance protein 1, Crkas, CRK-associated substrate, Crk-associated substrate p130Cas, FLJ12176, P130Cas | |
| BCAR1 | |
| IgG | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Human, Mouse, Rat | |
| 9564 | |
| p130 Cas (phospho Tyr410) antibody was raised against a peptide sequence around phosphorylation site of tyrosine 410 (G-V-Y (p)-A-V) derived from Human p130 Cas . | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title