missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p130Cas/BCAR1, p Tyr410 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54638-0.025mg
This item is not returnable.
View return policy
Description
p130Cas/BCAR1 Polyclonal specifically detects p130Cas/BCAR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| p130Cas/BCAR1 | |
| Polyclonal | |
| Unconjugated | |
| Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, and 50% glycerol. with 0.02% sodium azide | |
| BCAR1 | |
| p130 Cas (phospho Tyr410) antibody was raised against a peptide sequence around phosphorylation site of tyrosine 410 (G-V-Y (p)-A-V) derived from Human p130 Cas . | |
| 0.025 mg | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot, Immunohistochemistry | |
| 1 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100 | |
| breast cancer estrogen resistance 1, Cas scaffolding protein family member 1, CAS1, CASFLJ45059, CASS1CRKASbreast cancer estrogen resistance protein 1, Crkas, CRK-associated substrate, Crk-associated substrate p130Cas, FLJ12176, P130Cas | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9564 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction