Learn More
p130Cas Ab-1 Mouse Monoclonal Antibody, Epredia™
Take advantage of a quality product that delivers accurate results with the Epredia™ p130Cas Ab-1, Mouse Monoclonal Antibody.
£118.00 - £587.04
Specifications
| Antigen | p130superscriptCassuperscript Ab-1 |
|---|---|
| Clone | CAS-14; same as 35B-1A4 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Host Species | Mouse |
| Product Code | Brand | Quantity | Conjugate | Form | Concentration | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Conjugate | Form | Concentration | Price | Quantity & Availability | ||
|
12642447
|
Epredia
MS-855-B0 |
100 μL | Biotin | Concentrated | 200μg/mL | ||||
|
12672447
|
Epredia
MS-855-P0 |
100 μL | Unconjugated | Concentrated | 200μg/mL | ||||
|
12652447
|
Epredia
MS-855-B1 |
500 μL | Biotin | Concentrated | 200μg/mL | ||||
|
12692447
|
Epredia
MS-855-P1 |
500 μL | Unconjugated | Concentrated | 200μg/mL | ||||
|
12632447
|
Epredia
MS-855-B |
1 mL | Biotin | Concentrated | 200μg/mL | ||||
|
12662447
|
Epredia
MS-855-P |
1 mL | Unconjugated | Concentrated | 200μg/mL | ||||
|
12602457
|
Epredia
MS-855-P1ABX |
100 μL | Unconjugated | Concentrated | 1mg/mL | ||||
|
12682447
|
Epredia
MS-855-PABX |
200 μL | Unconjugated | Concentrated | 1mg/mL | ||||
|
12642457
|
Epredia
MS-855-R7 |
7 mL | Unconjugated | Prediluted | Prediluted | ||||
Descripción
The p130Cas (Cas for Crk-associated substrate) is a common cellular target of phosphorylation signal via v-Crk and v-Src oncoproteins. p130Cas has a unique structure that contains a Src homology (SH)-3 domain followed by multiple YXXP motifs and a proline-rich region p130Cas is implicated in a variety of biological processes including cell adhesion, cell migration, growth factor stimulation, cytokine receptor engagement and bacterial infection. Physiological functions of p130Cas include cardiovascular development, actin filament assembly and Src-induced cell transformation.Host Species: Mouse
Clone: CAS-14; same as 35B-1A4
Isotype: IgG1/κ
Species Reactivity: Human and Rat. Others not known.
Epitope: Not determined
Immunogen: Recombinant human p130Cas protein
Molecular Weight: 130kDa
Positive Control: MCF-7 Cells, HUVEC cells, Tonsil or breast CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Denatured verified)
Note:
USA: RUO; Int'l: RUO
Especificaciones
| p130superscriptCassuperscript Ab-1 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Mouse | |
| Cell Cycle | |
| Recombinant human p130superscriptCassuperscript protein | |
| Primary |
| CAS-14; same as 35B-1A4 | |
| Monoclonal | |
| RUO | |
| Human, Rat | |
| IgG2a κ | |
| MCF-7 cells, HUVEC cells, tonsil or breast CA |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.