Learn More
MEDICOM Weecover™ Max 1 Coveralls Type 5/6
Product Code. 31781427 Shop All Medicom Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31781427 Large Pack of 50
31781360 X-Large Pack of 50
31781419 2X-Large Pack of 50
31781375 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31781427 Supplier MEDICOM Supplier No. WLC103
  • ONLINE EXCLUSIVE PRICE
    £221.35
    £185.58 / Pack of 50
    Save £35.77
    16% Off
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
WEECOVER™ Max 1 Type 5/6 coverall provides reinforced protection against asbestos fibers while ensuring optimal comfort.
  • Flat thermosealed seams offering optimal sealing for asbestos fibers
  • Elastic waistband for a better fit
  • Special asbestos packaging for traceability of your PPE
  • Designed for maximum comfort with its ultra-breathable material
  • Loose fit to ensure the wearer freedom of movement and promote air circulation for better comfort
Specifications
Product Type Chemical Protective Clothing
Size Large
Gender Unisex
Material SMS Fabric
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Zipper
Seam Type Taped
Sterility Non-sterile
Resistance Chemicals, Liquid