Learn More
MEDICOM Weecover™ Max 1 Integral Coveralls Type 5/6
Product Code. 31781424 Shop All Medicom Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 20
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31781424 Large Pack of 20
31781376 X-Large Pack of 20
31781300 2X-Large Pack of 20
31781349 3X-Large Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31781424 Supplier MEDICOM Supplier No. WLC103I
  • ONLINE EXCLUSIVE PRICE
    £142.39
    £119.37 / Pack of 20
    Save £23.02
    16% Off
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
WEECOVER™ Max1 Integral Type 5/6 coverall combines optimal asbestos protection with comfort thanks to its ultra-breathable SMS fabric.
  • Raglan sleeves and loose fit for more comfort
  • Elasticated cuffs and ankles, elastic drawstring at the back
  • Flat thermobonded seams offering optimal protection against asbestos
  • Integrated overboots with non-slip sole
  • Elasticated thumb loops
Specifications
Product Type Chemical Protective Clothing
Size Large
Gender Unisex
Material SMS Fabric
Color White, Red
Disposable Single-use
Waist Style Standard
Ankle Style Attached Boots
Cuff Style Thumb Loop
Neck Style Hooded
Hood Type 3-panel
Closure Type Zipper
Seam Type Taped
Sterility Non-sterile
Resistance Chemicals, Liquid