Learn More
MEDICOM Weecover™ Coveralls Type 5/6
Product Code. 31781413 Shop All Medicom Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31781413 Medium Pack of 50
31781408 Large Pack of 50
31781291 X-Large Pack of 50
31781361 2X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31781413 Supplier MEDICOM Supplier No. WLC02
  • ONLINE EXCLUSIVE PRICE
    £151.41
    £126.95 / Pack of 50
    Save £24.46
    16% Off
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
WEECOVER™ Type 5/6 coverall is designed to provide maximum comfort thanks to its ultra-breathable SMS fabric and loose fit.
  • Designed for maximum comfort with its ultra-breathable material
  • Loose fit ensuring the wearer freedom of movement and promoting air circulation for better comfort
  • Elastics at the ankles and waist for a better fit
  • EN 1073-2 standard against radioactive particles
  • Jersey cuffs
Specifications
Product Type Chemical Protective Clothing
Size Medium
Gender Unisex
Material SMS Fabric
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Chemicals, Liquid