Learn More
MEDICOM Weepro™ Coveralls Type 5/6
Product Code. 31781395 Shop All Medicom Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
2X-Large
3X-Large
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31781395 Small Pack of 50
31781322 Medium Pack of 50
31781309 Large Pack of 50
31781383 X-Small Pack of 50
31781305 2X-Large Pack of 50
31781357 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31781395 Supplier MEDICOM Supplier No. WLP01
  • ONLINE EXCLUSIVE PRICE
    £176.36
    £147.87 / Pack of 50
    Save £28.49
    16% Off
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
WEEPRO™ Type 5/6 coverall offers breathable, comfortable protection, suitable for industrial environments and maintenance operations.
  • Breathable material for better comfort
  • Antistatic treated material facilitating the dissipation of electrostatic charges
  • Raglan sleeves and loose fit for better ease of movement
  • Elastics at the ankles and waist for a better fit
  • Jersey cuffs
Specifications
Product Type Chemical Protective Clothing
Size Small
Gender Unisex
Material Microporous Polyethylene Laminate (Non-woven)
Color White
Disposable Single-use
Waist Style Standard
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Chemicals, Liquid