Learn More
MEDICOM Weecover™ Blue Coveralls Type 5/6
Product Code. 31781389 Shop All Medicom Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31781389 Medium Pack of 50
31781329 Large Pack of 50
31781334 X-Large Pack of 50
31781351 2X-Large Pack of 50
31781292 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 31781389 Supplier MEDICOM Supplier No. WLCB02
  • ONLINE EXCLUSIVE PRICE
    £157.82
    £132.32 / Pack of 50
    Save £25.50
    16% Off
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
WEECOVER™ Blue Type 5/6 coverall offers comfort and breathability for dirty work, industrial maintenance and technical operations.
  • Designed for maximum comfort with its ultra-breathable material
  • Loose fit ensuring the wearer freedom of movement and promoting air circulation for better comfort
  • Elastics at the ankles and waist for a better fit
  • EN 1073-2 certified against radioactive particles
  • Jersey cuffs
  • Blue color for dirty work
Specifications
Product Type Chemical Protective Clothing
Size Medium
Gender Unisex
Material SMS Fabric
Color Blue
Disposable Single-use
Waist Style Standard
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Chemicals, Liquid