Learn More
MEDICOM Weecover™ Eco Coveralls Type 5/6
Product Code. 31781388 Shop All Medicom Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31781388 Large Pack of 50
31781407 X-Large Pack of 50
31781353 2X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31781388 Supplier MEDICOM Supplier No. WLCECO03
  • ONLINE EXCLUSIVE PRICE
    £138.66
    £116.26 / Pack of 50
    Save £22.40
    16% Off
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
WEECOVER™ Eco Type 5/6 coverall offers optimal comfort thanks to its ultra-breathable SMS fabric and is a cost-effective bulk solution.
  • Designed for maximum comfort with its ultra-breathable material
  • Loose fit for good freedom of movement
  • Elasticated ankles and waist for better fit
  • Jersey cuffs
  • No individual bag - 2 bags of 25 coveralls per carton
Specifications
Product Type Chemical Protective Clothing
Size Large
Gender Unisex
Material SMS Fabric
Color White
Disposable Single-use
Waist Style Standard
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Chemicals