Learn More
MEDICOM Hop'Safe Pro Coveralls Type 5/6
Product Code. 31781386 Shop All Medicom Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31781386 Large Pack of 25
31781425 X-Large Pack of 25
31781362 2X-Large Pack of 25
31781358 3X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31781386 Supplier MEDICOM Supplier No. 40.061/L
  • ONLINE EXCLUSIVE PRICE
    £228.33
    £191.43 / Pack of 25
    Save £36.90
    16% Off
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Hop'Safe Pro coverall provides breathable chemical protection with hood, sealed seams and reinforcements for demanding environments.
  • Hooded model with adhesive flap on zipper closure
  • Elastics at the wrists and ankles, hood and waist
  • Light and flexible
  • Knee reinforcement
  • Integrated thumb loops
  • Integrated boots
  • Permeable to air and water vapor
  • External seams for added comfort
  • Seams covered with adhesive for better protection against the penetration of liquid and solid chemicals as well as asbestos dust (compliant with the requirements of the decree of 7 March 2015)
  • The suit offers good tear and abrasion resistance
  • Individually packaged
Specifications
Product Type Hooded Coveralls with Attached Boots
Size Large
Gender Unisex
Material SMS Fabric
Color Red
Disposable Single-use
Waist Style Standard
Ankle Style Attached Boots
Cuff Style Thumb Loop
Neck Style Hooded
Hood Type 3-panel
Closure Type Zipper
Seam Type Sealed
Sterility Non-sterile
Resistance Chemicals, Liquid