Learn More
MEDICOM Weepro™ Max Green Coveralls Type 4/5/6
Product Code. 31781359 Shop All Medicom Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31781359 Medium Pack of 50
31781310 Large Pack of 50
31781422 X-Large Pack of 50
31781429 2X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31781359 Supplier MEDICOM Supplier No. WLPMG02
  • ONLINE EXCLUSIVE PRICE
    £363.67
    £304.91 / Pack of 50
    Save £58.76
    16% Off
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
WEEPRO™ MaxGreen Type 4/5/6 coverall provides liquid-tight protection against chemical splashes, ideal for crop protection treatments.
  • Thermosealed seams waterproof to liquid and aerosol splashes
  • Antistatic treated material facilitating the dissipation of electrostatic charges
  • Chinstrap for optimal fit around a full face respiratory mask
  • Elastics at wrists, ankles, and waist for a better fit
  • Ideal for phytosanitary treatments
Specifications
Product Type Chemical Protective Clothing
Size Medium
Gender Unisex
Material Microporous Polyethylene Laminate (Non-woven)
Color Green
Disposable Single-use
Waist Style Standard
Ankle Style Elastic
Cuff Style Thumb Loop
Neck Style Hooded
Hood Type 3-panel
Closure Type Zipper
Seam Type Taped
Sterility Non-sterile
Resistance Chemicals, Liquid