Learn More
MEDICOM Weejet™ Lite Coveralls Type 3/4/5/6
Product Code. 31781348 Shop All Medicom Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31781348 Medium Pack of 25
31781331 Large Pack of 25
31781341 X-Large Pack of 25
31781308 2X-Large Pack of 25
31781355 3X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 31781348 Supplier MEDICOM Supplier No. WLJ202
  • ONLINE EXCLUSIVE PRICE
    £229.73
    £192.60 / Pack of 25
    Save £37.13
    16% Off
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
WEEJET™ Lite coverall provides chemical protection Type 3/4/5/6 with a lightweight, silent fabric fully resistant to splashes.
  • Lightweight material, with a limited noise level, which remains sealed against liquid splashes
  • Increased protection on the front with its double adhesive flap and chin guard
  • Easy zip handling thanks to a ring that is easily accessible even with thick gloves
  • The seams with heat-sealed bands ensure perfect tightness
  • Protection against biological hazards and solid radioactive particles
Specifications
Product Type Chemical Protective Clothing
Size Medium
Gender Unisex
Material Polyethylene
Color Yellow
Disposable Single-use
Waist Style Standard
Ankle Style Elastic
Cuff Style Thumb Loop
Neck Style Hooded
Hood Type 3-panel
Closure Type Zipper
Seam Type Taped
Sterility Non-sterile
Resistance Chemicals, Liquid