Learn More
MEDICOM Weepro™ Max Integral Coveralls Type 4/5/6
Product Code. 31781345 Shop All Medicom Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
Unit Size:
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31781345 Small Pack of 50
31781369 Medium Pack of 50
31781290 Large Pack of 50
31781385 X-Small Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31781345 Supplier MEDICOM Supplier No. WLPMI01
  • ONLINE EXCLUSIVE PRICE
    £368.04
    £308.58 / Pack of 50
    Save £59.46
    16% Off
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
WEEPRO™ Max Integral Type 4/5/6 coverall offers reinforced liquid-tight protection against chemical and biological splashes, with integrated shoe covers.
  • Thermosealed seams waterproof to fine particle, liquid and aerosol splashes
  • Antistatic treated material facilitating the dissipation of electrostatic charges
  • Zipper closure with adhesive flap
  • Elastics at the wrists, ankles and waist for a better fit
  • Integrated overshoes
  • Tested against biological risks
Specifications
Product Type Chemical Protective Clothing
Size Small
Gender Unisex
Material Microporous Polyethylene Laminate (Non-woven)
Color White, Orange
Disposable Single-use
Waist Style Standard
Ankle Style Attached Boots
Cuff Style Thumb Loop
Neck Style Hooded
Hood Type 3-panel
Closure Type Zipper
Seam Type Taped
Sterility Non-sterile
Resistance Chemicals, Liquid