Learn More
MEDICOM Weepro™ Max+ Coveralls Type 4/5/6
Product Code. 31781336 Shop All Medicom Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31781336 Medium Pack of 50
31781352 Large Pack of 50
31781404 X-Large Pack of 50
31781367 2X-Large Pack of 50
31781307 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 31781336 Supplier MEDICOM Supplier No. WLPMP02
  • ONLINE EXCLUSIVE PRICE
    £286.17
    £239.93 / Pack of 50
    Save £46.24
    16% Off
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
WEEPRO™ Max+ Type 4/5/6 coverall is designed for high-risk environments, from asbestos removal to chemical and nuclear industries.
  • Heat-sealed seams resistant to liquid splashes and aerosols
  • Anti-static treated material to facilitate the dissipation of electrostatic charges
  • Elasticated wrists, ankles, and waist for better fit
  • Self-adhesive flap over the double zipper with chin guard for total seal with the mask
  • 3-panel hood for complete freedom of movement
  • Certified against radioactive particles
  • Certified against biological hazard
Specifications
Product Type Chemical Protective Clothing
Size Medium
Gender Unisex
Material Microporous Polyethylene Laminate (Non-woven)
Color White, Orange
Disposable Single-use
Waist Style Standard
Ankle Style Elastic
Cuff Style Thumb Loop
Neck Style Hooded
Hood Type 3-panel
Closure Type Zipper
Seam Type Taped
Sterility Non-sterile
Resistance Chemicals, Liquid