Learn More
MEDICOM Weeback™ Coveralls Type 5/6
Product Code. 31781316 Shop All Medicom Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31781316 Small Pack of 50
31781398 Medium Pack of 50
31781409 Large Pack of 50
31781297 X-Large Pack of 50
31781318 2X-Large Pack of 50
31781418 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31781316 Supplier MEDICOM Supplier No. WLB01
  • ONLINE EXCLUSIVE PRICE
    £268.90
    £225.45 / Pack of 50
    Save £43.45
    16% Off
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
WEEBACK™ Type 5/6 coverall provides waterproof splash protection while ensuring optimal comfort thanks to its SMS breathable back.
  • Thermally sealed seams waterproof to liquid and aerosol splashes
  • Gusseted crotch triangle for better durability
  • Elastic waist for a better fit
  • Significant mechanical resistance during intense work
  • Material treated with antistatic properties facilitating the dissipation of electrostatic charges
Specifications
Product Type Chemical Protective Clothing
Size Small
Gender Unisex
Material Microporous Polyethylene Laminate (Non-woven)
Color White, Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Zipper
Seam Type Taped
Sterility Non-sterile
Resistance Liquid