Learn More
Medicom WeePro Labo Integral Coverall with Boots, Type 5/6
Product Code. 17869291 Shop All Medicom Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 25
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17869291 Medium Pack of 25
17889291 Large Pack of 25
17809311 X-Large Pack of 25
17899291 2X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17869291 Supplier Medicom Supplier No. WLPLMI02
  • ONLINE EXCLUSIVE PRICE
    £233.24
    £206.40 / Pack of 25
    Save £26.84
    11% Off
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Coverall sterilized by gamma irradiation and cleanroom apparel compatible from ISO class 4.
  • New material MICROMIUM™, light and very low contaminant
  • Cleanroom apparel compatible from ISO class 4
  • Chemical splash suit, Category III Type 5/6
  • Double packaging fully sealed
  • Apparel sterilized by gamma irradiation
Specifications
Product Type Cleanroom Coveralls
Size Medium
Gender Unisex
Material Microporous Polyethylene Laminate
Color White
Disposable Single-use
Waist Style Elastic (Stitched-in)
Ankle Style Covered Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type 2-way Zipper
Seam Type Bound
Sterility Non-sterile
Resistance Chemicals
Latex Free No
No. of Pockets None
Cleanroom Class Class 10-100,000 (ISO 4-8)