Learn More
Medicom WeePro Collar Coverall, Type 5/6
Product Code. 17853745 Shop All Medicom Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17853745 Medium Pack of 50
17863745 Large Pack of 50
17873745 X-Large Pack of 50
17883745 2X-Large Pack of 50
17893745 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17853745 Supplier Medicom Supplier No. WLPC02
  • ONLINE EXCLUSIVE PRICE
    £214.07
    £184.35 / Pack of 50
    Save £29.72
    13% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Standardized against biological risks and radioactive particles, and constructed of breathable material for a better comfort.
  • Breathable material for a better comfort
  • Self-adhesive flap over zipper
  • Antistatic treatment to allow easy dissipation of electrostatic charges
  • Two-way zip closure
  • Elasticated ankles and waist for a better fit
  • Standardized against biological risks and radioactive particles
Specifications
Product Type Cleanroom Coveralls
Size Medium
Gender Unisex
Material Microporous Composite Fabric
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Knit
Neck Style Collared
Hood Type Unhooded
Closure Type 2-way Zipper
Seam Type Stitched
Sterility Non-sterile
Resistance Chemicals, Particle