Learn More
Medicom WeePro Labo Sterile Coverall, Type 5/6
Product Code. 17829301 Shop All Medicom Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 25
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17829301 Medium Pack of 25
17889301 Large Pack of 25
17839301 X-Large Pack of 25
17899301 2X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17829301 Supplier Medicom Supplier No. WLPLMS02
  • ONLINE EXCLUSIVE PRICE
    £248.15
    £220.22 / Pack of 25
    Save £27.93
    11% Off
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Coverall sterilized by gamma irradiation and cleanroom apparel compatible from ISO class 4.
  • New material MICROMIUM™, light and very low contaminant
  • Cleanroom apparel compatible from ISO class 4
  • Chemical splash suit, Category III Type 5/6
  • Double packaging fully sealed
  • Apparel sterilized by gamma irradiation
Specifications
Product Type Cleanroom Coveralls
Size Medium
Gender Unisex
Material Microporous Fabric
Color White
Disposable Single-use
Waist Style Elastic (Stitched-in)
Ankle Style Covered Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Zipper
Seam Type Bound
Sterility Sterile
Resistance Liquid
Latex Free No
No. of Pockets None
Cleanroom Class Class 10-100,000 (ISO 4-8)