missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf Research™ plus, Mechanical Single-Channel Pipettes
GREENER_CHOICE

Product Code. 15783439 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
0.1 to 2.5 μL
0.25 to 2.5 mL
0.5 to 10 μL
0.5 to 5 mL
1 to 10 mL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
Color:
Blue
Dark Gray
Light Gray
Medium Gray
Orange
Red
Turquoise
Violet
Yellow
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color unitSize
15783439 100 to 1000 μL Blue Each
15793439 0.5 to 5 mL Violet Each
15763439 10 to 100 μL Yellow Each
15733439 0.1 to 2.5 μL Dark Gray Each
15773439 20 to 200 μL Yellow Each
15753439 2 to 20 μL Yellow Each
15703449 1 to 10 mL Turquoise Each
15713449 2 to 20 μL Light Gray Each
15743439 0.5 to 10 μL Medium Gray Each
15723449 30 to 300 μL Orange Each
16262391 0.25 to 2.5 mL Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 15783439 Supplier Eppendorf Supplier No. 3123000063
Add to basket
Edge
Add to basket

Adapt for specific liquids or other external conditions with these ultra-light fully autoclavable pipettes. Pipettes feature a spring loaded tip cone, a secondary adjustment option and an improved volume display.

  • Volume adjustment feature allows desired volume to be reached in few quick turns
  • Control button and ejectors low operating force are positioned for ergonomics
  • 4-digit, magnifying-shape volume display
  • Calibration display and adjustment opening for specific liquids and volumes
  • Uses renowned PhysioCare Concept™ thus reduces strain on hand and arm during pipetting, maintains balance, minimal user exertion
  • Quick lock makes lower part particularly easy to remove and service (not available for 5 and 10 mL pipettes)
  • Spring loaded tip cone promotes tight fit to tip (not available for 5 and 10 mL pipettes)
  • Has ability to be autoclaved without disassembling

Applications

  • Pipetting liquids
  • Mixing liquids
  • Filling of plates, gels, and reaction vessels
  • Phase extraction and removal of supernatants
TRUSTED_SUSTAINABILITY

Specifications

Accuracy ±3%, ±1%, ±0.6%
Compatible Tips 1000 μL Pipette tips, 71mm L
Autoclavable Autoclavable
Color Blue
For Use With 1000 μL Pipette Tip
Number of Channels 1
Product Line Research Plus
Type Single-Channel Adjustable Volume Pipettes
Volume (Metric) 100 to 1000 μL
Display Digital
Finger Style Ergonomic
For Use With (Equipment) 1000 μL Pipette tip
Includes epT.I.P.S.™ Box 2.0 with 96 Pipette Tips
Increments 100, 500, 1000 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.