missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ School Grade Filter Paper
Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric):
90 mm
110 mm
180 mm
Unit Size:
Pack of 100
Description
- General purpose
- Available in a range of sizes
Specifications
Specifications
| Material | Cellulose |
| Grade | School grade |
| Quantity | 100 Pk. |
| Diameter (Metric) | 110 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction