Learn More
Fisherbrand™ MultiSUB Midi 96 Horizontal Gel System
Shop All Fisherbrand ProductsDescription
MultiSUB Horizontal Gel Electrophoresis Systems combine economical buffer and gel consumption without compromising resolution and separation speed. Leak-proof casting is simplified using casting dams, while the ultra-compact size minimizes bench space requirements.
The MultiSUB Midi 96 is ideal for high throughput electrophoresis of PCR products or samples containing a small number of DNA bands. The Midi 96 format can accommodate samples from a standard 96-well microplate, with direct loading using 8-channel pipettes. The offset layout of the wells allows a maximum run length of 1.8cm per well, resulting in improved separation.
- Designed for loading of DNA samples from multiwall plates
- Ideal for high throughput electrophoresis
- Average run-time is just 15 to 30 minutes
- Direct microplate format for easy lane identification
- Multichannel pipette compatible combs for speed loading
- Includes MultiSUB Midi96 tank/lid/electrode unit, 10 x 12cm UV tray, comb block (12 x 8-sample, 1mm thick combs), casting dams
Spécification
Spécification
| Color | Blue Lid, Clear Tank, White Block/Red Combs, Red/Black Electrodes |
| For Use With (Application) | High throughput electrophoresis |
| For Use With (Equipment) | 8 Channel Pipettes, 96-well Plates |
| Type | Horizontal Gel Electrophoresis System |
| Gel Dimensions | 10 x 12 cm |
| Includes | MultiSUB Midi96 tank/lid/electrode unit, 10 x 12cm UV tray, comb block (12 x 8-sample, 1mm thick combs), casting dams |
| Material | Injection-molded Plastic |
| No. of Wells | 96 samples plus 1 marker lane |
| Quantity | 1 Each |
| Thickness (Metric) | 1 mm (combs) |
| Afficher plus |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.