missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellROX™ Orange Reagent, for oxidative stress detection

Product Code. 12807322
Change view
Click to view available options
Quantity:
5 x 50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12807322 5 x 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Produktkod. 12807322 Leverantör Invitrogen™ Leverantörsnummer C10443

Please to purchase this item. Need a web account? Register with us today!

Edge
Denna artikel kan inte returneras. Se returpolicy

Innehåller

Five single-use vials with enough total reagent for five 96-well plates or 100 coverslips

CellROX™ Orange Reagent is a novel fluorogenic probe for measuring oxidative stress in live cells. This cell-permeant dye is non-fluorescent while in a reduced state and exhibits bright orange fluorescence upon oxidation by reactive oxygen species (ROS), with absorption/emission maxima of ∼545/565 nm.

CellROX™ Orange Reagent is a novel fluorogenic probe for measuring oxidative stress in live cells. This cell-permeant dye is non-fluorescent while in a reduced state and exhibits bright orange fluorescence upon oxidation by reactive oxygen species (ROS), with absorption/emission maxima of ∼545/565 nm. While this reagent is not formaldehyde-fixable, it can be multiplexed with other live cell-compatible probes such as GFP, CellEvent™ Caspase 3/7 Green reagent, Image-iT™ DEAD Green, Green and Deep Red MitoTracker™ dyes, and NucBlue™ reagent.

CellROX™ Orange Reagent is compatible with various platforms, such as traditional fluorescence microscopy, high content screening (HCS), and flow cytometery. This reagent is also compatible with the Tali™ Image-based Cytometer.

In addition, CellROX™ Orange Reagent is:
• Compatible with other live cell probes, including GFP, and may be added directly to serum-containing growth media
• Provided as a stable, ready-to-use DMSO solution with a simple protocol compatible with standard workflows in fluorescence microscopy
• Provided as five single-use vials containing enough total reagent for five 96-well plates or 100 coverslips

Oxidative stress results from an imbalance between the production of reactive oxygen species (ROS) and the ability of cells to scavenge them. ROS play an important role in the progression of several diseases, including inflammation, atherosclerosis, aging, and age-related degenerative disorders.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Beställningsinfo

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifikationer

Color Orange
Format Frozen
Quantity 5 x 50 μL
Content And Storage Store at ≤–20°C. Protect from light and desiccate.

For Use With (Equipment) Imaging, HCS, Cytometer
Concentration 2.5 mM stabilized solution in DMSO
Detection Method Live Cell Imaging
Excitation/Emission 545/565 nm
Indicator Oxidative stress
Product Line CellROX

For Research Use Only. Not for use in diagnostic procedures.

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.