missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ White Plastic Bio-Hazard Trays

Product Code. 12377207 Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W):
54 x 34 cm
57 x 54 cm
68 x 54 cm
70 x 46 cm
113 x 54 cm
Unit Size:
Each

Product Code. 12377207

Only null left
Add to basket
Add to basket

Available in general purpose, biohazard and radiation safety formats, spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection.

  • Available in general purpose, biohazard and radiation safety formats
  • Spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection
  • All trays supplied with one free liner when purchased
  • Additional environmentally friendly APET liners are provided in packs of 25

Specifications

Dimensions (L x W) 54 x 34 cm
Color White
Length (Metric) 54 cm
Material Plastic
Type Spill Containment Tray
Width (Metric) 34 cm
For Use With Safe containment of spillages, lab bench protection
Includes Liner
No. per Pack 1 Each
Shape Rectangular
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.