missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ BacLight™ RedoxSensor™ CTC Vitality Kit
BacLight™ RedoxSensor™ CTC Vitality Kit
Brand: Molecular Probes™ B34956
744.80 GBP valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00450kg
Description
The BacLight™ RedoxSensor™ CTC Vitality Kit provides effective reagents for evaluating bacterial cell health and vitality that can withstand fixation procedures. Briefly, healthy cells respiring via the electron transport chain will absorb and reduce CTC into an insoluble, red fluorescent formazan product. Cells not respiring or respiring at slower rates will reduce less CTC and consequently produce less fluorescent product, giving a semi-quantitative estimate of healthy vs. unhealthy bacteria. Green- and blue-fluorescent nucleic acid stains are included as counterstains to assist differentiation of cells from debris and calculating total cell numbers.Specifications
–5 to –30°C | |
Fluorescent | |
Flow Cytometer, Fluorescence Microscope | |
Slide(s), Tube(s) | |
1 Kit(s) |
BacLight™ RedoxSensor™ CTC Vitality Kit, 50 rxns, 350.25 Molecular Weight, C16H17Cl2N5 Molecular Formula, Fluorescent Detection, Compatible with Bacteria, –5 to –30°C Storage, Use with Flow Cytometer, Fluorescence Microscope | |
Cell Viability, Proliferation and Function | |
CTC: 450nm, DAPI: 344nm, SYTO™ 24: 490nm | |
Contains 5 vials of 5-cyano-2, 3-ditolyl tetrazolium chloride (CTC, 15mg per vial), 1 vial of SYTO™ 24 green (100μL in DMSO)m and DAPI (100μL, 5mg/mL solution in water) |
For Research Use Only. Not for use in diagnostic procedures.