missing translation for 'onlineSavingsMsg'
Learn More

P-3FAX-Neu5Ac, TRC

Product Code. 30391743
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391743 2.5 mg 2.5mg
30396932 5 mg -
30392063 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391743 Supplier Toronto Research Chemicals Supplier No. TRCF1010782.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117405-58-0
Molecular Formula C22H30FNO14
Molecular Weight (g/mol) 551.47
Synonym 5-(acetylamino)-3,5-dideoxy-3-fluoro-D-erythro-alpha-L-manno-2-nonulopyranosonic Acid Methyl Ester 2,4,7,8,9-Pentaacetate
SMILES COC(=O)C1(OC(=O)C)O[C@H]([C@H](OC(=O)C)[C@@H](COC(=O)C)OC(=O)C)[C@H](NC(=O)C)[C@@H](OC(=O)C)[C@H]1F

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C22H30FNO14/c1-9(25)24-16-18(17(35-12(4)28)15(34-11(3)27)8-33-10(2)26)38-22(21(31)32-7,37-14(6)30)20(23)19(16)36-13(5)29/h15-20H,8H2,1-7H3,(H,24,25)/t15-,16?,17-,18?,19?,20?,22?/m1/s1
Formula Weight 551.165
Chemical Name or Material P-3FAX-Neu5Ac
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.