CAS RN 117405-58-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ P-3FAX-Neu5Ac
Sialic acid analog; sialyltransferase inhibitor
2
P-3FAX-Neu5Ac, TRC
CAS: 117405-58-0 Molecular Formula: C22H30FNO14 Molecular Weight (g/mol): 551.47 Synonym: 5-(acetylamino)-3,5-dideoxy-3-fluoro-D-erythro-alpha-L-manno-2-nonulopyranosonic Acid Methyl Ester 2,4,7,8,9-Pentaacetate SMILES: COC(=O)C1(OC(=O)C)O[C@H]([C@H](OC(=O)C)[C@@H](COC(=O)C)OC(=O)C)[C@H](NC(=O)C)[C@@H](OC(=O)C)[C@H]1F